missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 6M, Honeywell™

Product Code. 15603220
Click to view available options
Quantity:
25 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 6 M HCl (6N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7647-01-0
Concentration 6M
Vapor Pressure 190mmHg (25°C)
Quantity 25 L
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
UN Number 1789
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric Acid Solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'