missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric Acid, TraceSELECT™, 34 to 37%, Honeywell™ Fluka™ Product Code.: 15676910

Hydrochloric Acid, TraceSELECT™, 34 to 37%, Honeywell™ Fluka™

Product Code. 15676910 Shop All Honeywell Fluka Products
2.5 L, Poly bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Poly bottle
This item is not returnable. View return policy

Product Code. 15676910

Brand: Honeywell-Fluka 727872.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TraceSELECT™, for inorganic trace analysis, 34-37%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Specifications

pH >0.1 (at 20°C)
Color Colorless
CAS 7647-01-0
Melting Point ∼-35°C
Boiling Point ∼42°C (1013hPa)
Density 1.2g/mL at 25°C
Odor Stinging
Packaging Poly bottle
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 2.5 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
UN Number UN1789
Beilstein 1098214
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Completely miscible in water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Percent Purity 34 to 37%
Grade TraceSELECT™
Chemical Name or Material Hydrochloric acid, For inorganic trace analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid, TraceSELECT™, 34 to 37%, Honeywell™ Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.