missing translation for 'onlineSavingsMsg'
Learn More

Hydrocinnamic acid, 99%

Product Code. 10193490 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10193490

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 501-52-0 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-52-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002771
InChI Key XMIIGOLPHOKFCH-UHFFFAOYSA-N
Synonym hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
PubChem CID 107
ChEBI CHEBI:28631
IUPAC Name 3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 47.0°C to 50.0°C
Density 1.0700g/mL
Boiling Point 279.0°C to 281.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2CO2H
Quantity 5 g
Beilstein 09, 508
Merck Index 15, 4822
Specific Gravity 1.07
Solubility Information Solubility in water: 0.5% in water; more soluble in hot water. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,benzene,ether,chloroform,,petroleum ether and carbon disulfide
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hydrocinnamic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.