missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 48%, Honeywell™ Fluka™

ACS Reagent, 48%

45.25 € - 67.51 €

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15608460
SDS View Documents
Honeywell-Fluka
695068-25ML
25 mL Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15636720
SDS View Documents
Honeywell-Fluka
695068-500ML
500 mL Poly bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7664-39-3
20.01
KRHYYFGTRYWZRS-UHFFFAOYSA-N
14917
hydrogen fluoride
FH
MFCD00011346
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
CHEBI:29228
F

Specifications

Acidic
7664-39-3
∼105°C
Stinging
FH
MFCD00011346
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
F
20.01
CHEBI:29228
48%
Colorless
-35°C
1.15g/mL at 25°C
25mmHg (20°C)
Liquid
UN1790B
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrogen fluoride
14917
20.01g/mol
Hydrofluoric acid
Videos
Safety and Handling

Safety and Handling

P260-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233

missing translation for 'dotInformation' : 6.1(8) / PGII

missing translation for 'rtecsNumber' : 231-634-8

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.