missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent

Specifications
Specifications
Color | Colorless |
Melting Point | -35.0°C |
Boiling Point | 105.0°C |
Density | 1.1500g/mL |
Heavy Metals (as Pb) | 0.5ppm max. |
Packaging | Plastic bottle |
Residue after Ignition | 0.0005% max. |
Specific Gravity | 1.15 |
Quantity | 500 g |
Linear Formula | HF |
Show More |
Safety and Handling


Product Identifier
- Hydrofluoric acid, 48 to 51% solution in water
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H310-Fatal in contact with skin.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H300-Fatal if swallowed.
- H330-Fatal if inhaled.
- H290-May be corrosive to metals.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction