missing translation for 'onlineSavingsMsg'
Learn More
Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals Product Code.: 11400191

Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals

Product Code. 11400191
500 mL
Click to view available options
Quantity:
250 mL
500 mL
1000 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11400191

Brand: Thermo Scientific Alfa Aesar 038746.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484Show MoreShow Less
PubChem CID 14917
ChEBI CHEBI:29228
SMILES F

Specifications

Color Colorless
Boiling Point 112°C
Density 1.16 g/mL
Odor Acrid
Quantity 500 mL
Physical Form Liquid, Double Sub-Boiling Quartz Distillation
UN Number UN1790
Merck Index 14,4790
IUPAC Name fluorane
Formula Weight 20.01
Percent Purity 47 to 51%
Grade Environmental Plus
Chemical Name or Material Hydrofluoric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS13J005 Certificate TypeCertificate of Analysis Date21/06/2023 Catalog Number038746,038746.AP
Lot NumberQ26I057 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number038746.AU,038746.AP,038746.AK
Lot NumberQ28I057 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number038746.AU,038746.AP,038746.AK
Lot NumberR03I035 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog Number038746.AU,038746.AP,038746.AK
Lot NumberQ28I014 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog Number038746.AU,038746.AP,038746.AK

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.