Learn More
hydrogen chloride, 4N solution in 1,4-dioxane, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 388361000
152.76 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Hydrochloric acid | |
4N solution in 1, 4-dioxane | |
ClH | |
MFCD00011324 MFCD00792839 | |
1.0500g/mL | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.45 | |
1.05 | |
17°C |
Colorless to White | |
7647-01-0, 123-91-1 | |
HCl | |
3.95 to 4.40N | |
14, 4794 | |
Solubility in water: soluble. Other solubilities: soluble in most organic solvents | |
Cl | |
36.46 | |
CHEBI:17883 | |
AcroSeal™ Glass Bottle | |
100 mL | |
Crystals or Crystalline Powder |
Safety and Handling
- Hydrogen chloride, 4N solution in 1,4-dioxane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H350-May cause cancer.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 1,4-Dioxane, Hydrochloric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only