Learn More
Hydrogen chloride, pure, 2N solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
111.00 € - 319.00 €
Specifications
Chemical Name or Material | Hydrochloric acid |
---|---|
Color | Colorless |
Name Note | 2N solution in diethyl ether |
CAS | 7647-01-0, 60-29-7 |
Concentration | 1.90 to 2.20N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10364552
|
Thermo Scientific
368471000 |
100 mL | AcroSeal™ Glass bottle |
111.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10782164
|
Thermo Scientific Acros
368478000 |
800 mL | AcroSeal™ Glass Bottle |
319.00 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydrochloric acid | |
2N solution in diethyl ether | |
1.90 to 2.20N | |
0.74 | |
ClH | |
Liquid | |
04,450; 05,533; 06,283 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
36.46 | |
CHEBI:17883 | |
Pure |
Colorless | |
7647-01-0, 60-29-7 | |
0.7400g/mL | |
−40°C | |
HCl | |
MFCD00011324 MFCD00792839 | |
10, 4701 | |
Solubility in water: soluble. Other solubilities: soluble in most organic solvents | |
Cl | |
313 | |
36.45 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Hydrogen chloride, 1N solution in diethyl ether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H224-Extremely flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only