missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, pure, 5 to 6N solution in 2-propanol

Product Code. p-4509480 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
Packaging:
Plastic bottle
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 10214950

Brand: Thermo Scientific Acros 133700010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrochloric acid
CAS 7647-01-0, 67-63-0
Color Colorless to Yellow
Physical Form Liquid
Quantity 1 L
Molecular Formula ClH
Linear Formula HCl
MDL Number MFCD00011324 MFCD00792839
Fieser 04,450; 05,533; 06,283
Flash Point 11°C
Merck Index 15,4831
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.45
Grade Pure
Concentration or Composition (by Analyte or Components) 5N min.
Packaging Plastic bottle
Specific Gravity 0.909
Density 0.9090g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen chloride, 5 to 6N solution in 2-propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.