missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen chloride solution, 4.0 M in 1,4-dioxane, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
800 mL
2 L
4 x 100 mL
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
UN Number | UN2924 |
Density | 1.05g/mL at 25°C |
IUPAC Name | chlorane |
Formula Weight | 36.46g/mol |
Packaging | Glass Bottle |
Quantity | 100 mL |
Flash Point | 17°C (62.6°F) |
Physical Form | Liquid |
Chemical Name or Material | Hydrogen chloride solution |
Safety and Handling
Product Identifier
- Hydrochloric acid in 1,4-Dioxane
- Danger
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 1,4-dioxane, hydrochloric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'