missing translation for 'onlineSavingsMsg'
Learn More

hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt, Thermo Scientific Chemicals

Product Code. 10114310
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym Hexachloroplatinic acid, Platinic chloride
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 60.0°C
Physical Form Chunks and/or Powder
Quantity 5 g
Linear Formula H2PtCl6·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 409.82
Percent Purity ∼40% (Pt)
Packaging Glass bottle
Chemical Name or Material Hydrogen hexachloroplatinate(IV) hydrate, ca. 40% Pt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'