missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 29-32%, (ACS Reagent Grade), MP Biomedicals
Click to view available options
Quantity:
500 mL
Specifications
Specifications
CAS | 7722-84-1 |
Quantity | 500 mL |
Assay Percent Range | 29-32% |
Molecular Formula | H2O2 |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
SMILES | OO |
IUPAC Name | hydrogen peroxide |
Molecular Weight (g/mol) | 34.014 |
PubChem CID | 784 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'