missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hydrogen peroxide, 3 wt.% solution in water, stabilized, Thermo Scientific Chemicals
25.73 € - 127.64 €
Chemical Identifiers
CAS | 7722-84-1, 7732-18-5 |
---|---|
Molecular Formula | H2O2 |
Molecular Weight (g/mol) | 34.014 |
MDL Number | MFCD00011333 |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
PubChem CID | 784 |
ChEBI | CHEBI:16240 |
IUPAC Name | hydrogen peroxide |
SMILES | OO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10081254
|
Thermo Scientific Acros
426000250 |
25 mL | Plastic bottle |
25.73 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10517824
|
Thermo Scientific Acros
426001000 |
100 mL | Plastic bottle |
44.42 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10061494
|
Thermo Scientific Acros
426000010 |
1 L | Plastic bottle |
127.64 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7722-84-1, 7732-18-5 | |
34.014 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
Specifications
7722-84-1, 7732-18-5 | |
H2O2 | |
MFCD00011333 | |
15, 4835 | |
Solubility in water: soluble. | |
OO | |
34.014 | |
CHEBI:16240 | |
1.01 |
100.0°C | |
HOOH | |
1.0100g/mL | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34 | |
Hydrogen peroxide, 3 wt.% solution in water, stabilized |
Safety and Handling
GHS H Statement:
Causes skin irritation.
RUO – Research Use Only