missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide 30-32% (w/w) (100 Volumes), Primar™, for Trace Metal Analysis, Fisher Chemical
Brand: Fisher Chemical H/1820/15
145.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
1L, HDPE plastic bottle

Specifications
Colorless | |
108°C | |
30-32%w/w | |
11333 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
HDPE plastic bottle | |
3.3 | |
Liquid |
7722-84-1 | |
-33°C | |
H2O2 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
1.1 | |
1 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrogen peroxide 100 volumes 30-32% w/w
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Hydrogen peroxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrogen Peroxide 30-32% (w/w) (100 Volumes), Primar™, for Trace Metal Analysis, Fisher Chemical > 1L, HDPE plastic bottle
Spot an opportunity for improvement?Share a Content Correction