missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide 6% (w/v) (20 Volumes), Extra Pure SLR, Fisher Chemical™

Product Code. 10274050
Click to view available options
Quantity:
1 L
2.5 L

Hydrogen Peroxide 6% (w/v) (20 Volumes), Extra Pure SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number 11333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 100°C
Melting Point 0°C
Formula Weight 34.0128g/mol
Packaging HDPE plastic bottle
pH 2.5 to 3.5
Quantity 2.5 L
Physical Form Liquid
compliance-icons
Product Identifier
  • Hydrogen peroxide 20 volumes >6% w/v
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen peroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'