missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide Solution, ACS Reagent, Honeywell™
ACS Reagent, contains inhibitor, 30 wt. % in H2O
34.45 € - 501.10 €
Chemical Identifiers
CAS | 7722-84-1 |
---|---|
Molecular Formula | H2O2 |
Molecular Weight (g/mol) | 34.014 |
MDL Number | MFCD00011333 |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
PubChem CID | 784 |
ChEBI | CHEBI:16240 |
IUPAC Name | hydrogen peroxide |
SMILES | OO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15632040
|
Honeywell Research Chemicals
216763-100ML |
100 mL |
34.45 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15642040
|
Honeywell Research Chemicals
216763-500ML |
500 mL |
84.14 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15658330
|
Honeywell Research Chemicals
216763-4L |
4 L |
501.10 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
7722-84-1 | |
34.014 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
Specifications
Undesignated | |
H2O2 | |
2014 | |
30wt. % in H2O | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34.01g/mol | |
23.3mmHg (30°C) | |
Hydrogen Peroxide Solution, Contains inhibitor |
7722-84-1 | |
MFCD00011333 | |
3587191 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
ACS Reagent | |
1 (vs air) |
Safety and Handling
P280-P305 + P351 + P338 + P310
H318-H412
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrogen peroxide solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : hydrogen peroxide solution