missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, Stabilized, 0.3, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Pressure Bottom Poly Bottle
Specifications
Specifications
Quantity | 500 mL |
Formula Weight | 34.01 |
Percent Purity | 30% |
Grade | Meets ACS reagent chemical requirements |
Packaging | HDPE Pressure Bottom Poly Bottle |
Specific Gravity | 1L = 1.11kg |
Chemical Name or Material | Hydrogen Peroxide, Stabilized |
Safety and Handling


Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrogen Peroxide, Stabilized, 0.3, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction