missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Hydrogen Peroxide Cell Test Kit

With certificate for optimum precision.

Brand:  WTW™ 250402


Product Code. 11784006

  • 188.00 € / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For infrequent or very large sample throughput in the laboratory
  • Sample adding, inserting, measuring and reading
  • Reaction cuvette with barcode or method selection

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Copper reaction with phenanthroline derivate
PhotoLab™ S12, PhotoLab™ Spectral photometers
25
25 Tests
Hydrogen peroxide (H2O2) cell test kit
For waste water, drinking water, disinfectants, detergents
Certificates
0.25 - 20.0mg/I H₂O₂
Photometric
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrogen Peroxide Cell Test Method

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

Supplemental information
  • MIXTURE LIST-Contains : methanol, sulphuricacid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers