missing translation for 'onlineSavingsMsg'
Learn More
Hydrogen tetrabromoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 32% min, Thermo Scientific Chemicals Product Code.: 11334038

Hydrogen tetrabromoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 32% min, Thermo Scientific Chemicals

Product Code. 11334038
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11334038

Brand: Thermo Scientific Alfa Aesar 040433.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Au{C(NHMe) 2 } 2 ]Cl·H 2 O and [Au{C(NHMe) 2 } 2 ]Br·H 2 O were obtained by adding methyl isocyanide and 40% aqueous methylamine successively to aqueous solutions of hydrogen tetrachloroaurate(III) hydrate and hydrogen tetrabromoaurate(III) hydrate, respectively. To analyze the influence of the bromide ions in the synthesis of the gold nanorods Hydrogen tetrabromoaurate(III) hydrate is used. Hydrogen tetrabromoaurate(III) hydrate was used for purification in continuous microfluidic synthesis and functionalization of gold nanorods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00054118
Synonym Bromoauric acid

Specifications

Melting Point ∼27°C
Physical Form Crystalline
Quantity 5 g
UN Number UN3260
Solubility Information Soluble in water and alcohol
Formula Weight 517.61 (Anhydrous)
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Au 32% min.
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Hydrogen tetrabromoaurate(III) hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN02I035 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040433.06,040433.03,040434.06
Lot NumberB04Q07 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040431.06,040433.06,040431.03
Lot NumberK05M10 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040431.06,040433.06,040431.03
Lot NumberB15L11 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040431.06,040433.06,040431.03
Lot NumberD22M03 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040433.06,040433.03,040434.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrogen tetrabromoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 32% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.