Learn More
Hydrogen tetrachloroaurate(III) hydrate, Premion™, 99.995% (metals basis), Au 49% min, Thermo Scientific Chemicals
Description
Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Crystalline |
Quantity | 1 g |
Assay Percent Range | 99.999% (metals basis), Au 49% min |
Solubility Information | Soluble in water. Insoluble in ether. |
Formula Weight | 339.79 (anhydrous) |
Sensitivity | Hygroscopic; Light sensitive |
Chemical Name or Material | Hydrogen tetrachloroaurate(III) hydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.