missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetrachloroaurate(III) hydrate, Premion™, 99.995% (metals basis), Au 49% min, Thermo Scientific Chemicals

200.00 € - 820.08 €

Chemical Identifiers

CAS 27988-77-8
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149903
InChI Key RFWCEGCWRSNAGT-UHFFFAOYSA-K
IUPAC Name hydrogen tetrachlorogoldtris(ylium)
SMILES [H+].Cl[Au+3](Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11345627
SDS View Documents
Thermo Scientific Alfa Aesar
042803.03
1 g
200.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11335627
SDS View Documents
Thermo Scientific Alfa Aesar
042803.06
5 g
820.08 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27988-77-8
339.77
RFWCEGCWRSNAGT-UHFFFAOYSA-K
[H+].Cl[Au+3](Cl)(Cl)Cl
AuCl4H
MFCD00149903
hydrogen tetrachlorogoldtris(ylium)

Specifications

27988-77-8
99.999% (metals basis), Au 49% min
MFCD00149903
RFWCEGCWRSNAGT-UHFFFAOYSA-K
hydrogen tetrachlorogoldtris(ylium)
339.79 (anhydrous)
Hydrogen tetrachloroaurate(III) hydrate
Crystalline
AuCl4H
Soluble in water. Insoluble in ether.
[H+].Cl[Au+3](Cl)(Cl)Cl
339.77
Hygroscopic; Light sensitive
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302-H314-H317-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 240-948-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.