missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetranitratoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 38.9-40.1%, Thermo Scientific Chemicals

Product Code. 11384028
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for Isomerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00046178
Synonym Nitratoauric acid

Specifications

Melting Point ∼72°C (decomposition)
Physical Form Crystalline
Quantity 5 g
UN Number UN1477
Solubility Information Decomposes in water; Soluble in HNO3
Formula Weight 445.99 (Anhydrous)
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Au 38.9 to 40.1%
Assay (metals basis)
Chemical Name or Material Hydrogen tetranitratoaurate(III) hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.