Learn More
VEE GEE Hydrometer Jar
The Hydrometer Jar is made of borosilicate glass, the jar features a beaded rim and a circular base. Sized properly to eliminate surface tension effects that lead to inaccurate readings.
Brand:  VEE GEE 21050-500
 View more versions of this product
Product Code. 17816583
  • 20.30 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The capacity varies depending on the model.
Specifications
Specifications
Flat Circular
14 in.
Hydrometer Jar
500 mL
Reusable
17 fl. oz.
36 cm
10.8 cm
Borosilicate glass