Learn More
Fisherbrand™ Hydrometer Rack, 10-Place
GREENER_CHOICE
Product Code. p-200084620 Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Gray
Unit Size:
Each
Product Code. 30130292
Only null left
Add to basket
Edge
Add to basket
Rack for gentle handling and strong protection for fragile glass hydrometers.
The Hydrometer Rack accommodates up to 10 hydrometers with diameters up to 31 mm, ensuring safe storage, efficient drying, and secure transport.
Secure Organization
  • Teardrop-Shaped Slots hold hydrometers securely in place, support a range of diameters, and ensure easy insertion and removal for convenient handling
  • 3-Tier Setup provides enhanced stability and protection of hydrometer body
Clean and Convenient
  • Built-in Handles provide a secure grip for safe and convenient transport
  • Removable Drip Tray simplifies cleaning and maintenance, improving sanitation and protecting your workspace
Robust Design
  • Stable Wide Base ensures increased stability and secure placement for hydrometers
  • Durable Polypropylene Construction is easy to clean, long-lasting, and fully recyclable
Specifications
Includes Drip Tray
Product Type Hydrometer Rack
Autoclavable Not Autoclavable
Color Gray
Holds Hydrometers (Long), Hydrometers (Short)
Material Polypropylene
Positions 10
Shelves 3 Tiers
Size 29.5 x 17.50 x 23.40 cm
Length (Metric) 29.5 cm
Width (Metric) 17.50 cm
Height (Metric) 23.4 cm