Learn More
Brannan™ Hydrometer specific gravity shot loaded low adjustment for surface tension 0.950 to 1.000 x 0.001 S.G. 270mm Brannan
Brand:  Brannan™ 10/923/0
Product Code. 12801468
  • 17.50 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Hydrometer specific gravity shot loaded low adjustment for surface tension 0.950 to 1.000 x 0.001 S.G. 270mm