Learn More
Brannan™ Hydrometer specific gravity shot loaded medium adjustment for surface tension 1.300 to 1.350 x 0.001 S.G. 270 mm Brannan
Slot loaded density hydrometer for a variety of surface tension ranges.
Brand:  Brannan™ 10/930/0
Product Code. 12871468
  • 17.50 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Specific Gravity Hydrometer
270 mm
Soda Glass
0.001 g/mL
1.30 to 1.35 g/mL
Steel Shot