Learn More
Brannan™ Hydrometer specific gravity shot loaded medium adjustment for surface tension 1.400 to 1.450 x 0.001 S.G. 270 mm Brannan
Slot loaded density hydrometer for a variety of surface tension ranges.
Brand:  Brannan™ 10/932/0
Product Code. 12891468
  • 21.05 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Specific Gravity Hydrometer
270 mm
Soda Glass
0.001 g/mL
1.40 to 1.45 g/mL
Steel Shot