missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone, 99.5%, Thermo Scientific Chemicals

26.23 € - 269.34 €

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00002339
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10313292
View Documents
Thermo Scientific Acros
219930050
5 g Glass Bottle
27.24 €
5g
Please to purchase this item. Need a web account? Register with us today!
10012840
View Documents
Thermo Scientific Acros
219930500
50 g Plastic bottle
26.23 €
50g
Please to purchase this item. Need a web account? Register with us today!
10454715
View Documents
Thermo Scientific Acros
219932500
250 g Plastic bottle
36.88 €
250g
Please to purchase this item. Need a web account? Register with us today!
10761335
View Documents
Thermo Scientific Acros
219930010
1 kg Plastic bottle
90.75 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15149253
View Documents
Thermo Scientific Acros
219930051
5 kg Plastic Drum
269.34 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-31-9
110.11
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
785
benzene-1,4-diol
C6H6O2
MFCD00002339
hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
CHEBI:17594
C1=CC(=CC=C1O)O

Specifications

123-31-9
White
285.0°C to 287.0°C
Authentic
C6H6O2
MFCD00002339
05,341; 14,249
hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
benzene-1,4-diol
785
110.11
Needle-like Crystals or Crystalline Powder
170.0°C to 174.0°C
1.32
165°C
99.4% min. (HPLC)
C6H4(OH)2
06,836
15,4845
Solubility in water: 70g/L in water (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,readily soluble in ethanol,acetone and methanol
C1=CC(=CC=C1O)O
110.11
CHEBI:17594
99.5%
Hydroquinone, p.a.
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Suspected of causing cancer.
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-617-8

missing translation for 'rtecsNumber' : MX3500000

missing translation for 'tsca' : TSCA

Produktidentifikator
  • Hydroquinone

Signalwort
  • Danger

Gefahrenkategorie
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1

Gefahrenhinweis
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.

Sicherheitshinweis
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt