missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxychloroquine sulfate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Hydroxychloroquine sulfate shows antimalarial activity against malaria parasites mainly in the intra-erythrocytic phase of its life cycle. It acts by inhibiting the heme polymerase inside the food vacuole of the parasites
- It shows an inhibitory activity in autophagy preventing the lysosome fusion with the autophagosome
- Some studies demonstrated that hydroxychloroquine sulfate, due to its antiviral activity can restrict HIV infection in vitro
Applications
- Hydroxychloroquine sulfate displays antiviral and anti-rheumatic activities
- In vitro and in vivo experiments have been performed to investigate the role of hydroxychloroquine sulfate in the treatment of SARS-CoV-2 disease
- Hydroxychloroquine sulfate can be used in immunofluorescence assays to evaluate the antiviral mechanisms in dengue virus-2 infection. In this disease, this compound can activate the innate immune response
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Melting Point | 240.0°C |
Color | White |
Loss on Drying | 2% max. (105°C, 2 hrs.) |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.98 |
Packaging | Glass bottle |
Quantity | 25 g |
Merck Index | 15, 4857 |
Solubility Information | Solubility in water: soluble. Other solubilities: practically insoluble in alcohol,chloroform,,ether,acetone and acetic acid |
Formula Weight | 433.96 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0420952 | Certificate of Analysis | 16/01/2023 | 263010250,263011000 |
A0401377 | Certificate of Analysis | 15/12/2022 | 263010250,263011000 |
A0370447 | Certificate of Analysis | 15/12/2022 | 263010250,263011000 |
A0261705 | Certificate of Analysis | 15/12/2020 | |
A0324051 | Certificate of Analysis | 15/12/2020 |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Hydroxychloroquine sulfate
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydroxychloroquine sulphate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydroxychloroquine sulfate, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction