missing translation for 'onlineSavingsMsg'
Learn More
Hydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 11458880

Hydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11458880
100 g
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11458880

Brand: Thermo Scientific Alfa Aesar A15398.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.

Solubility
Soluble in water, alcohol, methanol and glycerol.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, phosphorous pentachloride, calcium and anhydrous copper(II) sulfate.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine;hydrochloride
SMILES Cl.NO

Especificaciones

Melting Point 152°C (dec.)
pH 3.6
Quantity 100 g
Linear Formula NH2OH·HCl
UN Number UN2923
Beilstein 3539763
Merck Index 14,4828
Solubility Information Soluble in water,alcohol,methanol and glycerol.
Formula Weight 69.49
Percent Purity 99%
Odor Acidic
Sensitivity Hygroscopic
Density 1.67
Chemical Name or Material Hydroxylamine hydrochloride
Mostrar más Mostrar menos

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253277 Certificate TypeCertificate of Analysis Date29/11/2024 Catalog NumberA15398.0I,A15398.30,A15398.22,A15398.0B,A15398.36
Lot Number10250138 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog NumberA15398.0I,A15398.30,A15398.22,A15398.0B,A15398.36
Lot Number10247741 Certificate TypeCertificate of Analysis Date18/08/2023 Catalog NumberA15398.0I,A15398.30,A15398.22,A15398.0B,A15398.36
Lot NumberW15I001 Certificate TypeCertificate of Analysis Date23/09/2022 Catalog NumberA15398.0I,A15398.30,A15398.22,A15398.0B,A15398.36
Lot Number5005H24T Certificate TypeCertificate of Analysis Date18/08/2022 Catalog NumberA15398.0I,A15398.30,A15398.22,A15398.0B
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Hydroxylamine hydrochloride
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Indicaciones de peligro
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
Consejos de prudencia
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Otros peligros
  • MIXTURE LIST-Contains : Hydroxylamine, hydrochloride

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Hydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado