missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine hydrochloride, 99+%, Thermo Scientific Chemicals

37.29 € - 805.01 €

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
SMILES Cl.NO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10442441
View Documents Promotion Details
Thermo Scientific Acros
270101000
100 g Plastic bottle
37.29 €
100g
Please to purchase this item. Need a web account? Register with us today!
10551121
View Documents Promotion Details
Thermo Scientific Acros
270102500
250 g Plastic bottle
56.58 €
250g
Please to purchase this item. Need a web account? Register with us today!
10164490
View Documents Promotion Details
Thermo Scientific Acros
270100010
1 kg Plastic bottle
197.99 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10593484
View Documents Promotion Details
Thermo Scientific Acros
270100050
5 kg Plastic drum
805.01 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

5470-11-1
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
443297
Cl.NO
ClH4NO
MFCD00051089
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
CHEBI:5807

Specifications

155°C to 158°C
White
99+%
H2NOH·HCl
01,478; 02,217; 07,176; 09,245
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine;hydrochloride
443297
69.49
Authentic
5470-11-1
Crystalline Powder, Crystals and/or Chunks
ClH4NO
MFCD00051089
15, 4867
Solubility in water: 560g/L (20°C). Other solubilities: soluble in alcohol,methanol,glycerol,propylene glycol,insoluble in cold ether
Cl.NO
69.49
CHEBI:5807
≥99%
Hydroxylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing canc

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protecti

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 226-798-2

Product Identifier
  • Hydroxylamine hydrochloride

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only