missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine-O-sulfonic acid, 97%, Thermo Scientific Chemicals

Product Code. 10692322
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2950-43-8
Molecular Formula H3NO4S
Molecular Weight (g/mol) 113.09
MDL Number MFCD00011604
InChI Key DQPBABKTKYNPMH-UHFFFAOYSA-N
Synonym hydroxylamine-o-sulfonic acid, aminooxy sulfonic acid, sulfoperamidic acid, hydroxylamine-o-sulphonic acid, permonosulfamic acid, amidoperoxymonosulfuric acid, aminooxysulfonic acid, amidosulfonic peracid, haos, sulfamic acid n-oxide
PubChem CID 76284
IUPAC Name amino hydrogen sulfate
SMILES NOS(O)(=O)=O

Specifications

Melting Point 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Linear Formula H2NOSO3H
Quantity 100 g
Fieser 01,481; 02,217; 03,156; 04,256; 05,343; 06,290; 08,250; 09,245; 10,207; 15,17
Solubility Information Solubility in water: soluble
Formula Weight 113.09
Percent Purity 0.97
Physical Form Crystalline Powder
Chemical Name or Material Hydroxylamine-O-sulfonic acid, 0.97
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydroxylamine-O-sulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'