missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylapatite, for analysis, Thermo Scientific Chemicals

Product Code. 10110273
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-06-5
Molecular Formula Ca5HO13P3
Molecular Weight (g/mol) 502.31
MDL Number MFCD00010904
InChI Key XYJRXVWERLGGKC-UHFFFAOYSA-D
Synonym Calcium phosphate hydroxide
IUPAC Name pentacalcium hydroxide triphosphate
SMILES [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Melting Point 1700.0°C
Color White
Physical Form Powder
Quantity 1 kg
Linear Formula 3Ca3(PO4)2·Ca(OH)2
Merck Index 15, 3514
Solubility Information Solubility in water: negligible
Formula Weight 502.31
Percent Purity 35 to 40% (Ca)
Grade Analytical
Loss on Ignition 8% max. (800°C)
Packaging Plastic Bottle
Chemical Name or Material Hydroxylapatite
Show More Show Less
Product Identifier
  • Hydroxylapatite

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'