missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Hydroxypropyl cellulose, Average M.W. 100.000
80.80 € - 1153.74 €
Chemical Identifiers
CAS | 9004-64-2 |
---|---|
Molecular Formula | (C24H44O16)n |
MDL Number | MFCD00132688 |
Synonym | 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10723191
|
Thermo Scientific Chemicals
184881000 |
100 g | Glass bottle |
80.80 €
100g |
In Stock
Log in to see stock. |
||||
10477380
|
Thermo Scientific Chemicals
184882500 |
250 g | Glass bottle |
154.77 €
250g |
In Stock
Log in to see stock. |
||||
10667753
|
Thermo Scientific Chemicals
184880010 |
1 kg | Plastic bottle |
368.84 €
1kg |
In Stock
Log in to see stock. |
||||
15129223
|
Thermo Scientific Chemicals
184880051 |
5 kg | Plastic drum |
1153.74 €
5kg |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9004-64-2 | |
(C24H44O16)n | |
MFCD00132688 | |
4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c |
Specifications
White to Cream | |
9004-64-2 | |
(C24H44O16)n | |
4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c | |
Authentic | |
75-150 mPa.s 5% aq.sol. |
5 to 8.5 (1% soln.) | |
Granules or Crystalline Powder | |
MFCD00132688 | |
100.000 (Average ) | |
5% max. (105°C) | |
Hydroxypropyl cellulose |
Safety and Handling
missing translation for 'rtecsNumber' : NF9050000
missing translation for 'tsca' : TSCA
Product Identifier
- Hydroxypropyl cellulose, Average M.W. 100.000
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title