Learn More
Hydroxyurea, 98%, Thermo Scientific Chemicals
Description
Hydroxyurea is used as an inhibitor of DNA synthesis and an anti-cancer drug. It is used as a precursor to prepare 1-hydroxy-5,6-dimethyl-1H-pyrimidine-2,4-dione via reaction with 2-methyl-3-oxo-butyric acid ethyl ester. It is used in the treatment of melanoma, resistant chronic myelocytic leukemia and recurrent metastatic or inoperable ovarian. It is also involved in reducing the rate of painful attacks in sickle-cell disease and has anti-retroviral properties in diseases such as AIDS.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water, hot alcohol and dimethyl sulfoxide. Insoluble in ethanol and benzene.
Notes
Moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible

Specifications
Specifications
Melting Point | ∼140°C (decomposition) |
Quantity | 25 g |
Beilstein | 1741548 |
Merck Index | 14,4848 |
Solubility Information | Soluble in water,hot alcohol and dimethyl sulfoxide. Insoluble in ethanol and benzene. |
Formula Weight | 76.06 |
Percent Purity | 98% |
Chemical Name or Material | Hydroxyurea |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W01J007 | Certificate of Analysis | 21/09/2023 | C22322.30 |
10245808 | Certificate of Analysis | 09/05/2023 | C22322.30 |
10207101 | Certificate of Analysis | 22/11/2021 | C22322.30 |
10190485 | Certificate of Analysis | 22/11/2021 | C22322.30 |
10233087 | Certificate of Analysis | 22/11/2021 | C22322.30 |
Safety and Handling

- Hydroxyurea
- Danger
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1A
- H340-May cause genetic defects.
- H360D-May damage the unborn child.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydroxyurea
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.