missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ HyFlex™ 11-280 Sleeves
GREENER_CHOICE

Product Code. 17677455
Click to view available options
Size:
Narrow
Wide
This item is not returnable. View return policy
This item is not returnable. View return policy

Ansell™ HyFlex™ 11-280 Sleeves feature a seamless design and options of lengths and widths to ensure an excellent fit for all-day comfort

  • INTERCEPT™ Cut Resistance Technology provides best-in-class cut protection and a soft and cool feel to the worker
  • Engineered to perform in combination with any HyFlex cut resistant glove
  • Available in three versions: Extra Wide, Wide, Narrow
TRUSTED_SUSTAINABILITY

Specifications

Size Wide
Construction Knitted
Product Type Sleeve
Material HPPE Yarn
Color Gray
Sterility Non-sterile
Disposable Reusable
Model HyFlex™11-280
For Use With (Application) Glass repair and replacement; Handling metal sheets and panels
Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category ll
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'