missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-500 Medium-Duty Industrial Gloves

Industrial gloves offer comfort and a sure grip. Ideal for dry or slightly oil applications where workers need light cut protection.

Brand:  Ansell 11500070

132.38 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 11746414

  • 150.00 € / 12 pairs
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for assembly and maintenance operations involving handling of sharp-edged and pointed tools and materials
  • Cut-resistant stretch Kevlar™ liner provides comfortable fit and protection from nuisance nicks and abrasion
  • Black foam nitrile coating on palm and fingertips channels oil away from glove surface
  • Extended cuff for extra cut protection around wrist
  • Yellow
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI 3
Knit
Nitrile Foam
ANSI A2
Cut-resistant Gloves
EN 388:3231B, EN 388:3231
Knit
HyFlex™ 11-500
19.8 cm
CE 0493, EN 420:2003 + A1:2009; Category III
Nitrile
Black, Yellow
Yes
7
Automotive, machinery and equipment, metal fabrication
Nylon, Lycra™, Aramid Fiber
7.8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.