missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-600W White Nylon Gloves with Polyurethane Coating

Product Code. 12610712
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Polyurethane coating on the palm improves tactile sensitivity, while the white color palette is useful for clean applications.

  • Designed for light and delicate applications in dry environments
  • Bare-handed dexterity with protection for light duty applications
  • Polyurethane coating on the palm enhances tactile sensitivity
  • White color is ideal for clean applications
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Cuff Material Polyurethane
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A1
Hand Specific
Material Nylon
Size 6
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 11-600W
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.