missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-605 Light Duty Nylon Gloves with Polyurethane Coated Fingertips

Product Code. 12620722
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12620722

Brand: Ansell 11605070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Industrial gloves designed for light and delicate applications in dry environments that require high levels of dexterity.

  • Nylon gloves are comfortable and flexible
  • Polyurethane coating on the fingertips enhances tactile sensitivity
  • Designed for light and delicate applications that require high levels of dexterity
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Fingertips
Coating Material Polyurethane
Color White, Yellow
Cut Resistance Rating ANSI A1
Exterior Finish Fingertip Textured
Hand Specific
Latex Free Yes
Material Nylon
Product Type Dip-coated Gloves
Size 7
EN388 Standard 003XA
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 11-605
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.