Learn More
Ansell™ HyFlex™ 11-616 Series Black and Green/Blue Polyurethane Light Weight Gloves
Product Code. 12600732
Click to view available options
Size:
6
7
8
9
10
11
Designed for a wide range of lightweight applications in dry or slightly oily environments. Offers outstanding comfort, fit and sensitivity for the most delicate handling.
  • 18-gauge knitting technology allows this glove to be ultra-thin yet tough
  • Ideal for workers who need a higher level of fit, touch and precision
  • Ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, EN 388 : 3121
Cuff Material Polyurethane
Coating Material Polyurethane
Product Type Lightweight Gloves
Size 9
Style 11-616
Glove Cuff Style Knit
Lining Nylon
Palm Type Coated-palm
Quantity 144 Pairs
Series 11-616