Learn More
Ansell Edmont™ HyFlex™ 11-625 Series White Polyurethane Medium Weight Gloves
Product Code. 12303579
Click to view available options
Size:
6
7
8
9
10
Deliver a perfect balance of comfort and sure handling with superior cut resistance. Innovative yarn structure contains an optimal rate of Dyneema™ offering high levels of cut resistance for a safe use in controlling, handling and assembly applications involving contacts with sharp pieces.
  • Perfect balance of comfort and sure handling with superior cut resistance.
  • The liners include an innovative yarn structure that contains an optimal rate of Dyneema™.
  • Offers high levels of cut resistance
  • The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
  • Designed for use in dry, as well as slightly oily environments.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, Standard CATEGORY II EN 388 : 4342
Cuff Material Polyurethane
Product Type Medium Weight Gloves
Size 9
Glove Cuff Style Knitwrist
Liner Material Dyneema™
Lining Spandex/Dyneema™
Palm Type Coated-palm
Quantity 12 Pairs
Resistant To High cut resistance