missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-651 Medium Duty Cut-Resistant Industrial Gloves

Product Code. 16019641
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Intercept™ Cut Resistance Technology blends engineered and synthetic fibers into high performance yarns to provide cut protection, along with comfort and dexterity.

  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Constructed with a seamless, 13-gauge liner
  • Elastic knit wrist ensures comfort, dexterity and a secure fit
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Cuff Material Elastic
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A3
Hand Specific
Material Intercept Technology Yarn
Size 7
EN388 Standard 4X42C
For Use With (Application) Machinery, Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-651
Yarn Gauge 13
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'