missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell HyFlex™ 11-751 Medium Duty Abrasion-Resistant Gloves, INTERCEPT™ Technology
Click to view available options
Size:
6
7
8
9
10
11
Description
- INTERCEPT Technology™ for high level of cut protection
- High durability PU coating, increased levels of abrasion resistance
- Dirt masking design allows for extended product use
- In compliance with the standards EN420 (General requirements) and EN388 (Mechanical hazards)
- Also available in vendor pack
Specifications
Specifications
ANSI Abrasion Rating | ANSI 6 |
Certifications/Compliance (Europe) | CE, EN 420:2003 + A1:2009; Category II |
Construction | Knit |
Coating Material | Polyurethane |
Color | Black |
Cut Resistance Rating | ANSI A4 |
Hand | Specific |
Latex Free | Yes |
Product Type | Cut-resistant Gloves |
Size | 10 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction