missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-819 Nylon/Spandex ESD Gloves with FORTIX™ Nitrile Foam Coating
GREENER_CHOICE

Product Code. 17200167
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Safety gloves with ESD functionality certified to EN 16350:2014, protecting products against electrostatic discharge. Ultra-thin FORTIX™ coating enhances grip, while also maintaining bare hand sensitivity for tasks that require dexterity and precision.

  • ESD functionality certified to EN 16350:2014
  • Can be used in explosion hazard zones as part of a grounding chain
  • Seamless, 18 gauge nylon and spandex liner helps maintain a bare-handed feel for tasks that require dexterity and precision
  • FORTIX™ nitrile foam coating enhances grip and provides long lasting protection
  • ZONZ™ Comfort Fit Technology ensures gloves are ultra breathable
  • Proprietary washing process helps to deliver cleaner, more comfortable gloves
  • Touchscreen functionality allows users to work with multiple touchscreen devices without glove removal
  • Dermatologically approved by Dermatest™
  • Certified by OEKO-TEX™
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 16350:2014, EN 420:2003 + A1:2009
Coated Area Palm
Applications Industrial
Coating Material Nitrile Foam
Color Blue
Disposable Reusable
Hand Specific
Latex Free Yes
Material Nylon, Spandex
Product Type ESD Gloves
Size 9
Sterility Non-sterile
EN388 Standard 3121A
For Use With (Application) Aerospace, Automotive, Logistics, and Machinery/Equipment
Glove Cuff Style Knit
Palm Type Coated
Series HyFlex™ 11-819
Yarn Gauge 18
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'