missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-931 Cut-Resistant Lightweight Gloves, Palm Coated
GREENER_CHOICE

Lightest weight cut-resistant and oil-repellent gloves with reinforced thumb crotch that provides up to 12x durability for extended wear.

243.00 €

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category II
Construction Knit
Coating Material Nitrile Foam
Color Black
View More Specs

Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16089691
GREENER_CHOICE
View Documents
Ansell
11931060
6
243.00 €
12 pairs
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
 
16099691
GREENER_CHOICE
View Documents
Ansell
11931070
7
243.00 €
12 pairs
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
16009701
GREENER_CHOICE
View Documents
Ansell
11931080
8
243.00 €
12 pairs
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
16019701
GREENER_CHOICE
View Documents
Ansell
11931090
9
243.00 €
12 pairs
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
16029701
GREENER_CHOICE
View Documents
Ansell
11931100
10
243.00 €
12 pairs
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
 
16039701
GREENER_CHOICE
View Documents
Ansell
11931110
11
243.00 €
12 pairs
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
 
Description

Description

  • RIPEL™ technology keeps oil from making contact with skin, keeping hands safe and dry
  • Provides protection from cuts and oil exposure, from light to heavy, across multiple applications
  • Provides dexterity and high level of comfort with ZONZ technology
  • ANSELL GRIP™ Technology for safe handling in dry-to-oily applications
  • FORTIX Technology for extended product use life and increased protection with up to 12x durability in thumb crotch
  • Ultra-lightweight 18 gauge design provides dexterity and high level of comfort
  • Protects against lacerations, oil and industrial fluid exposure
Specifications

Specifications

ANSI 4
Knit
Black
Cut-resistant Gloves
Inspection, assembly, fastening components, equipment repair and maintenance, automotive applications, press shop operations
Dyneema™ Diamond Technology
HyFlex™ 11-391
8.3 in.
CE, EN 420:2003 + A1:2009; Category II
Nitrile Foam
ANSI A2
EN 388:4X42B
Ansell GRIP™ Technology
ISO 188889
Reinforced
21.2 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.