missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-600W White Nylon Gloves with Polyurethane Coating

Product Code. 12690702
Click to view available options
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Polyurethane coating on the palm improves tactile sensitivity, while the white color palette is useful for clean applications.

  • Designed for light and delicate applications in dry environments
  • Bare-handed dexterity with protection for light duty applications
  • Polyurethane coating on the palm enhances tactile sensitivity
  • White color is ideal for clean applications
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A1
Hand Specific
Material Nylon
Size 10
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 11-600W
Yarn Gauge 15
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'