Learn More
3M™ Clean-Trace™ Hygiene Monitoring and Management System LM1 ATP System
Simple, quick and reliable hand held diagnostic tool to verify cleaning effectiveness and capture, store and manage data for later analysis.
Brand: 3M™ Clean-Trace™ 7100067976
This item is not returnable.
View return policy
Description
- Re-engineered for improved precision and accuracy
- Easy to use and fast time to result
- Robust for the manufacturing environment
- System includes the 3M™ Clean-Trace™ Luminometer and 3M™ Clean-Trace™ Hygiene Monitoring and Management Software to help you monitor testing results over time
- Ergonomic design for simple one-handed operation
- Wireless data transfer
- 23 languages for multilingual pre-production teams.
- Robust to provide consistent results across temperatures.
- New software has easy-to-use dashboard that immediately turns data into actionable information.
Specifications
Instrument | |
3 M™ Clean-Trace™ Luminometer, 3 M™ Clean-Trace™ Hygiene Monitoring and Management Software |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Clean-Trace™ Hygiene Monitoring and Management System LM1 ATP System
Spot an opportunity for improvement?Share a Content Correction