Learn More
Sampling Systems™ Hygienic Butterfly Valve Kit
These hygienic butterfly valves are manufactured from 316L stainless steel and are ideal for many applications, including GMP use.
259.00 € - 262.00 €
Specifications
Product Type Valve Kit
For Use With (Application) Dispensing Liquid
For Use With (Equipment) Drum with Sloping Base
Sterility Non-sterile
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17857817
View Documents
Sampling Systems™
A136-01A
Valve with EPDM Seals
262.00 €
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17912102
View Documents
Sampling Systems™
A136-01S
Valve with Silicone Seals
259.00 €
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
Description
Description
  • Comes with a silicone or EPDM seal
  • Both seals are suitable for food and pharmaceutical use and come with relevant certification
  • Easy to operate: Simply turn the handle to open or close
  • Opening the handle slightly will mean you have reduced product flow
  • Valves are ideal for liquids
Specifications
Specifications
Valve Kit
Dispensing Liquid
Drum with Sloping Base
Non-sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sampling Systems™ Hygienic Butterfly Valve Kit