Learn More
Ansell™ Hynit™ 32-105 Series Blue Nitrile Medium Weight Gloves
Special nitrile formulation unites excellent resistance to abrasion with remarkable flexibility. Liquid-repellent impregnation repels oil, grease and dirt.
46.50 €
Specifications
Back Material Nitrile
Certifications/Compliance CE Mark, ANSI, EN 388 : 3111
Cuff Material Nitrile
Coating Material Nitrile
Style 32-105
View More Specs
Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12680822
View Documents
Ansell™
32105070
7
46.50 €
12 pairs
In Stock
to see stock.
Edge
Add to basket
12690822
View Documents
Ansell™
32105075
7.5
46.50 €
12 pairs
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
Description
Description
Ideal for general plant use, assembly inspection, receiving, and light to medium fabrication and materials handling.
  • Nitrile-impregnated fabric ensures excellent abrasion and snag resistance
  • Feature clute cut and interlock knit liner for superior dry grip and excellent dexterity
  • Gloves are liquid repellent and machine washable so they will not shrink, stiffen, or lose their grip
  • Lining will not ball up
  • Color-coded cuffs for easy size identification and size mating after laundering
  • Available with solid and air-conditioned backs
Specifications
Specifications
Nitrile
Nitrile
32-105
Interlock Knit Polyester Cotton
144 Pairs
32-105
CE Mark, ANSI, EN 388 : 3111
Nitrile
Slip-on
Fully coated-palm
Excellent snag, puncture, abrasion and cut resistance