Learn More
Ansell™ Hynit™ 32-815 Series Blue Nitrile Light Weight Gloves Product Code.: 12660842
Add to basket
Ansell™ Hynit™ 32-815 Series Blue Nitrile Light Weight Gloves
Product Code. 12660842 Shop All Ansell Products
9
Click to view available options
Size:
7
7.5
8
9
10
Product Code. 12660842
Brand: Ansell™ 32815090
Edge
Add to basket
Add to basket
Special nitrile formulation unites excellent resistance to abrasion with remarkable flexibility. Liquid-repellent impregnation repels oil, grease and dirt.
  • Cut and sewn construction
  • Nitrile coated
  • Polyester cotton liner
  • Blue and white color
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance CE Mark, ANSI, EN 388 : 3111
Cuff Material Nitrile
Coating Material Nitrile
Product Type Lightweight Gloves
Size 9
Style 32-815
Glove Cuff Style Safety
Lining Interlock Knit Polyester Cotton
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Excellent snag, puncture, abrasion and cut resistance
Series 32-815
Product Title
Ansell™ Hynit™ 32-815 Series Blue Nitrile Light Weight Gloves >